missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1-(4-Nitrophenyl)azepane, 97%

Brand:  Alfa Aesar™ H63887.03

Additional Details : CAS : 13663-23-5 Weight : 0.00100kg Transport : UN number : 3259 Chem class : 8 Pack group : III

Quantity 1g

 View more versions of this product

Product Code. 15440839

  • € 59.00 / 1g


Estimated Shipment date
from Supplier 03-07-2020
Add to basket

Specifications

Chemical Identifiers

13663-23-5
220.272
WXAAQKMTSQDMII-UHFFFAOYSA-N
83639
C1CCCN(CC1)C2=CC=C(C=C2)[N+](=O)[O-]
C12H16N2O2
MFCD00156364
1-4-nitrophenyl azepane, 1h-azepine, hexahydro-1-4-nitrophenyl, 1-4-nitro-phenyl-azepane, n-4'-nitrophenyl hexamethylenimine, hexahydro-1-4-nitrophenyl-1h-azepine, 4-nitrophenyl azaperhydroepine, 1-4-nitrophenyl-perhydroazepin, 1-4-nitrophenyl hexahydro-1h-azepine
1-(4-nitrophenyl)azepane

Specifications

1-(4-Nitrophenyl)azepane
13663-23-5
MFCD00156364
1-4-nitrophenyl azepane, 1h-azepine, hexahydro-1-4-nitrophenyl, 1-4-nitro-phenyl-azepane, n-4'-nitrophenyl hexamethylenimine, hexahydro-1-4-nitrophenyl-1h-azepine, 4-nitrophenyl azaperhydroepine, 1-4-nitrophenyl-perhydroazepin, 1-4-nitrophenyl hexahydro-1h-azepine
C1CCCN(CC1)C2=CC=C(C=C2)[N+](=O)[O-]
220.272
220.27
97%
1g
C12H16N2O2
UN3259
WXAAQKMTSQDMII-UHFFFAOYSA-N
1-(4-nitrophenyl)azepane
83639
Solid
Videos
Documents