Learn More
1,6-Hexanediol bis[3-(3,5-di-tert-butyl-4-hydroxyphenyl)propionate], TRC
Specifications
Specifications
| Chemical Name or Material | Irganox 259 |
| CAS | 35074-77-2 |
| Molecular Formula | C40 H62 O6 |
| Quantity | 50 mg |
| Synonym | Benzenepropanoic acid, 3,5-bis(1,1-dimethylethyl)-4-hydroxy-, 1,1'-(1,6-hexanediyl) ester, Benzenepropanoic acid, 3,5-bis(1,1-dimethylethyl)-4-hydroxy-, 1,6-hexanediyl ester (9CI), 1,6-Bis[3-(3,5-di-tert-butyl-4-hydroxyphenyl)propionyloxy]hexane, 1,6-Bis[β-(3,5-di-tert-butyl-4-hydroxyphenyl)propionyloxy]hexane, 1,6-Hexamethylene bis(3,5-di-tert-butyl-4-hydroxyhydrocinnamate), 1,6-Hexanediol bis[3-(3,5-di-tert-butyl-4-hydroxyphenyl)propionate], 1,6-Hexanediol bis[β-(3,5-di-tert-butyl-4-hydroxyphenyl)propionate], 1,6-Hexanediyl-bis(3,5-bis(1,1-dimethylethyl)-4-hydroxybenzenepropanoate), Antioxidant 259, Hexamethylene 3-(3,5-di-tert-butyl-4-hydroxyphenyl)propionate, Hexamethylene bis(3,5-di-tert-butyl-4-hydroxyhydrocinnamate), Hexamethylene bis[3-(3,5-di-tert-butyl-4-hydroxyphenyl)propionate], Hexamethylene bis[β-(3,5-di-tert-butyl-4-hydroxyphenyl)propionate], Hexamethylene glycol bis(3,5-di-tert-butyl-4-hydroxyhydrocinnamate), Hexamethylene glycol bis[3-(3,5-di-tert-butyl-4-hydroxyphenyl)propionate], Irganox 249, Irganox 259 |
| InChI Formula | InChI=1S/C40H62O6/c1-37(2,3)29-23-27(24-30(35(29)43)38(4,5)6)17-19-33(41)45-21-15-13-14-16-22-46-34(42)20-18-28-25-31(39(7,8)9)36(44)32(26-28)40(10,11)12/h23-26,43-44H,13-22H2,1-12H3 |
| SMILES | CC(C)(C)c1cc(CCC(=O)OCCCCCCOC(=O)CCc2cc(c(O)c(c2)C(C)(C)C)C(C)(C)C)cc(c1O)C(C)(C)C |
| IUPAC Name | 6-[3-(3,5-ditert-butyl-4-hydroxyphenyl)propanoyloxy]hexyl 3-(3,5-ditert-butyl-4-hydroxyphenyl)propanoate |
| Molecular Weight (g/mol) | 638.92 |
| Formula Weight | 638.4546 |