Learn More
1,9-Nonanediol, 97%
Product Code. 11302476
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
Product Code. 11302476
Brand: Thermo Scientific Alfa Aesar L06336.14
Request Bulk or Custom Format
CAS: 3937-56-2 | C9H20O2 | 160.257 g/mol
1,9-Nonanediol is used in organic synthesis. It also can react with 3,4-dihydro-2H-pyran and 9-(tetrahydropyrane-2-yloxy)nonan-1-ol.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1,9-Nonanediol is used in organic synthesis. It also can react with 3,4-dihydro-2H-pyran and 9-(tetrahydropyrane-2-yloxy)nonan-1-ol.
Solubility
Soluble in methanol.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are Strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 3937-56-2
Molecular Formula C9H20O2
Molecular Weight (g/mol) 160.257
MDL Number MFCD00002991
InChI Key ALVZNPYWJMLXKV-UHFFFAOYSA-N
Synonym 1,9-nonanediol, 1,9-dihydroxynonane, .alpha.,.omega.-nonanediol, nonamethylene glycol, alpha,omega-nonanediol, 1,9-nonandiol, 1,9-nonane-diol, nonan-1,9-diol, acmc-209j4x
PubChem CID 19835
IUPAC Name nonane-1,9-diol
SMILES C(CCCCO)CCCCO
Specifications
Melting Point 45°C to 48°C
Density 0.95
Boiling Point 288°C
Flash Point 162°C (323°F)
Quantity 25 g
Beilstein 1737531
Solubility Information Soluble in methanol.
Formula Weight 160.26
Percent Purity 97%
Chemical Name or Material 1,9-Nonanediol