Learn More
1-Acetamidonaphthalene, TRC
Specifications
Specifications
| Chemical Name or Material | 1-Acetamidonaphthalene |
| CAS | 575-36-0 |
| Molecular Formula | C12 H11 N O |
| Quantity | 5 g |
| Synonym | Acetamide, N-1-naphthyl- (6CI,7CI,8CI), N-1-Naphthalenylacetamide, 1-(Acetylamino)naphthalene, 1-Acetamidonaphthalene, N-1-Naphthylacetamide, N-Acetyl-1-naphthalenamine, N-Acetyl-1-naphthylamine, N-Acetyl-α-naphthylamine, N-α-Naphthylacetamide, NSC 3105 |
| InChI Formula | InChI=1S/C12H11NO/c1-9(14)13-12-8-4-6-10-5-2-3-7-11(10)12/h2-8H,1H3,(H,13,14) |
| SMILES | CC(=O)Nc1cccc2ccccc12 |
| IUPAC Name | N-naphthalen-1-ylacetamide |
| Molecular Weight (g/mol) | 185.22 |
| Formula Weight | 185.0841 |