Learn More
1-Acetylazepan-2-one, 98%, BLD Pharm™
Specifications
Specifications
| CAS | 1888-91-1 |
| Molecular Formula | C8H13NO2 |
| MDL Number | MFCD00003262 |
| Quantity | 5 g |
| SMILES | O=C1N(C(C)=O)CCCCC1 |
| Molecular Weight (g/mol) | 155.19 |
| Percent Purity | 98% |
| Chemical Name or Material | 1-Acetylazepan-2-one |