Learn More
1-Benzyl-4-Boc-piperazine, 99%, Thermo Scientific Chemicals
Catalog Number 11363626
Click to view available options
Quantity:
1 g
5 g
57260-70-5
C16H24N2O2
276.38
MFCD00075603
GVHSMUYEAWMYLM-UHFFFAOYSA-N
1-benzyl-4-boc-piperazine, 1-boc-4-benzyl piperazine, 1-boc-4-benzyl-piperazine, tert-butyl-4-benzyl-1-piperazine carboxylate, 1-piperazinecarboxylic acid, 4-phenylmethyl-, 1,1-dimethylethyl ester, 4-benzyl-piperazine-1-carboxylic acid tert-butyl ester, maybridge4_002104, pubchem8596, 1-boc-4-benzylpiperazine, acmc-1bz2p
584330
tert-butyl 4-benzylpiperazine-1-carboxylate
CC(C)(C)OC(=O)N1CCN(CC2=CC=CC=C2)CC1
57260-70-5
C16H24N2O2
276.38
MFCD00075603
GVHSMUYEAWMYLM-UHFFFAOYSA-N
1-benzyl-4-boc-piperazine, 1-boc-4-benzyl piperazine, 1-boc-4-benzyl-piperazine, tert-butyl-4-benzyl-1-piperazine carboxylate, 1-piperazinecarboxylic acid, 4-phenylmethyl-, 1,1-dimethylethyl ester, 4-benzyl-piperazine-1-carboxylic acid tert-butyl ester, maybridge4_002104, pubchem8596, 1-boc-4-benzylpiperazine, acmc-1bz2p
584330
tert-butyl 4-benzylpiperazine-1-carboxylate
CC(C)(C)OC(=O)N1CCN(CC2=CC=CC=C2)CC1
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Melting Point 71°C to 73°C
Quantity 1 g
Beilstein 797033
Formula Weight 276.38
Percent Purity 99%
Chemical Name or Material 1-Benzyl-4-Boc-piperazine