missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1-Benzyl-5-cyanoindole, 97%

Brand:  Alfa Aesar™ H63188.06

Additional Details : CAS : 80531-13-1 Weight : 0.00500kg Transport : UN number : 3439 Chem class : 6.1 Pack group : III

Quantity 5g

 View more versions of this product

Product Code. 15439748

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Specifications

Chemical Identifiers

80531-13-1
232.286
XZCSETGZSGBGCI-UHFFFAOYSA-N
16114683
C1=CC=C(C=C1)CN2C=CC3=C2C=CC(=C3)C#N
C16H12N2
MFCD15200964
1-benzyl-1h-indole-5-carbonitrile, 1h-indole-5-carbonitrile, 1-phenylmethyl
1-benzylindole-5-carbonitrile

Specifications

1-Benzyl-5-cyanoindole
5g
C16H12N2
UN3439
1-benzyl-1h-indole-5-carbonitrile, 1h-indole-5-carbonitrile, 1-phenylmethyl
C1=CC=C(C=C1)CN2C=CC3=C2C=CC(=C3)C#N
232.286
232.28
97%
White to Brown
80531-13-1
MFCD15200964
6209162
XZCSETGZSGBGCI-UHFFFAOYSA-N
1-benzylindole-5-carbonitrile
16114683
Solid
Videos
Documents