Learn More
1-((Benzyloxy)methyl)cyclopropanecarbaldehyde, TRC
Specifications
Specifications
| Chemical Name or Material | 1-((Benzyloxy)methyl)cyclopropanecarbaldehyde |
| CAS | 288401-39-8 |
| Molecular Formula | C12H14O2 |
| Quantity | 2.5 g |
| InChI Formula | InChI=1S/C12H14O2/c13-9-12(6-7-12)10-14-8-11-4-2-1-3-5-11/h1-5,9H,6-8,10H2 |
| SMILES | O=CC1(COCc2ccccc2)CC1 |
| IUPAC Name | 1-(phenylmethoxymethyl)cyclopropane-1-carbaldehyde |
| Molecular Weight (g/mol) | 190.24 |
| Formula Weight | 190.0994 |