missing translation for 'onlineSavingsMsg'
Learn More

1-Boc-5-(tert-butyldimethylsiloxy)indole-2-boronic acid, 98%, Thermo Scientific™

Catalog Number p-7034812
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
335649-61-1
C19H30BNO5Si
391.346
MFCD08689532
NXKIUYOUKLIWLP-UHFFFAOYSA-N
5-tert-butyldimethylsilyloxy-1h-indole-2-boronic acid, n-boc protected, 1-boc-5-tbdmso-indole-2-boronic acid, 1-tert-butoxycarbonyl-5-tert-butyldimethylsilyloxy-1h-indol-2-boronic acid, 1-tert-butoxycarbonyl-5-tert-butyldimethylsilyloxy-1h-indol-2-ylboronic acid, 1-tert-butoxycarbonyl-5-tert-butyl dimethyl silyl oxy-1h-indol-2-yl boronic acid, 1-tert-butoxycarbonyl-5-tert-butyldimethylsilyl oxy indol-2-ylboronic acid, 1-boc-5-tbdmso-2-indoleboronic acid, 1-boc-5-tert-butyldimethylsiloxy indole-2-boronic acid, 1-tert-butoxycarbonyl-5-tert-butyldimethylsilyloxy indole, 5-tert-butyldimethylsilyloxy-1h-indole-2-boronic acid,n-boc protected 98
22285134
[5-[tert-butyl(dimethyl)silyl]oxy-1-[(2-methylpropan-2-yl)oxycarbonyl]indol-2-yl]boronic acid
B(C1=CC2=C(N1C(=O)OC(C)(C)C)C=CC(=C2)O[Si](C)(C)C(C)(C)C)(O)O
This item is not returnable. View return policy
335649-61-1
C19H30BNO5Si
391.346
MFCD08689532
NXKIUYOUKLIWLP-UHFFFAOYSA-N
5-tert-butyldimethylsilyloxy-1h-indole-2-boronic acid, n-boc protected, 1-boc-5-tbdmso-indole-2-boronic acid, 1-tert-butoxycarbonyl-5-tert-butyldimethylsilyloxy-1h-indol-2-boronic acid, 1-tert-butoxycarbonyl-5-tert-butyldimethylsilyloxy-1h-indol-2-ylboronic acid, 1-tert-butoxycarbonyl-5-tert-butyl dimethyl silyl oxy-1h-indol-2-yl boronic acid, 1-tert-butoxycarbonyl-5-tert-butyldimethylsilyl oxy indol-2-ylboronic acid, 1-boc-5-tbdmso-2-indoleboronic acid, 1-boc-5-tert-butyldimethylsiloxy indole-2-boronic acid, 1-tert-butoxycarbonyl-5-tert-butyldimethylsilyloxy indole, 5-tert-butyldimethylsilyloxy-1h-indole-2-boronic acid,n-boc protected 98
22285134
[5-[tert-butyl(dimethyl)silyl]oxy-1-[(2-methylpropan-2-yl)oxycarbonyl]indol-2-yl]boronic acid
B(C1=CC2=C(N1C(=O)OC(C)(C)C)C=CC(=C2)O[Si](C)(C)C(C)(C)C)(O)O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 104°C to 106°C
Quantity 250 mg
Formula Weight 391.35
Percent Purity 98%
Chemical Name or Material 1-Boc-5-(tert-butyldimethylsiloxy)indole-2-boronic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'