Learn More
1-Bromo-2-methoxy-d3-ethane, CDN
Product Code. 30316814
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.1g
0.25g
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30316814
Brand: CDN CDND78630.25G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 1-Bromo-2-methoxyethane-D3 (methoxy-D3)
CAS 2870708-27-1
Molecular Formula C3 D3 H4 Br O
Quantity 0.25 g
Synonym 1-Bromo-2-methoxy-d3-ethane, 1-Bromo-2-(methoxy-d3)ethane, 1-Bromo-2-methoxyethane-D3 (methoxy-D3), 2-Bromoethyl Methyl-d3 Ether
InChI Formula InChI=1 S/C3H7BrO/c1-5-3-2-4/h2-3H2,1H3/i1D3
SMILES [2 H]C([2 H])([2 H])OCCBr
IUPAC Name 1-bromo-2-(trideuteriomethoxy)ethane
Molecular Weight (g/mol) 142.0096
Formula Weight 140.9869 g/mol
Percent Purity 99 atom % D, min 98% Chemical Purity