missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-3-isopropylbenzene, 95%, Thermo Scientific™

Product Code. p-7047118
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11348596 1 g 1g
11358596 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11348596 Supplier Thermo Scientific Alfa Aesar Supplier No. L17654.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

Solubility
Difficult to mix in water.

Notes
Store in cool dry conditions in well sealed containers. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5433-01-2
MDL Number MFCD01318112

Specifications

Density 1.285
Boiling Point 210°C
Molecular Formula C9H11Br
Refractive Index 1.534
Quantity 1 g
Beilstein 1858092
Synonym 1-bromo-3-isopropylbenzene, 3-isopropylbromobenzene, 3-bromocumene, 3-bromoisopropylbenzene, benzene, 1-bromo-3-1-methylethyl, 1-bromo-3-isopropyl-benzene, 1-bromo-3-1-methylethyl benzene, m-bromocumene, m-isopropylbromobenzene, m-bromo-isopropyl-benzene
Solubility Information Difficult to mix in water.
InChI Key GBSGGFCCQZUXNB-UHFFFAOYSA-N
SMILES CC(C)C1=CC(=CC=C1)Br
IUPAC Name 1-bromo-3-propan-2-ylbenzene
Molecular Weight (g/mol) 199.09
PubChem CID 138499
Formula Weight 199.1
Percent Purity 95%
Chemical Name or Material 1-Bromo-3-isopropylbenzene
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.