Learn More
1-Bromo-4-chloro-2-nitrobenzene, 98%
Product Code. 11490473
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
Product Code. 11490473
Brand: Thermo Scientific Alfa Aesar A12635.14
Request Bulk or Custom Format
CAS: 41513-04-6 | C6H3BrClNO2 | 236.449 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 41513-04-6
Molecular Formula C6H3BrClNO2
Molecular Weight (g/mol) 236.449
MDL Number MFCD00024320
InChI Key UKTIMFAJRPSNGR-UHFFFAOYSA-N
Synonym 2-bromo-5-chloronitrobenzene, benzene, 1-bromo-4-chloro-2-nitro, 4-bromo-3-nitrochlorobenzene, 3-chloro-6-bromonitrobenzene, pubchem3593, acmc-209jkn, 1-bromo-4-chloronitrobenzene, ksc490m7r, 2-bromo-5-chloro-nitrobenzene, bromo-4-chloro-2-nitrobenzene
PubChem CID 2794904
IUPAC Name 1-bromo-4-chloro-2-nitrobenzene
SMILES C1=CC(=C(C=C1Cl)[N+](=O)[O-])Br
Specifications
Melting Point 68°C to 72°C
Quantity 25 g
Beilstein 1950424
Formula Weight 236.45
Percent Purity 98%
Chemical Name or Material 1-Bromo-4-chloro-2-nitrobenzene