Learn More
1-Bromobenzene, TRC
Click to view available options
Quantity:
5 g
Unit Size:
5g
Specifications
Specifications
| Chemical Name or Material | Bromobenzene |
| CAS | 108-86-1 |
| Purity Grade Notes | GC |
| Molecular Formula | C6 H5 Br |
| Quantity | 5 g |
| Synonym | Bromobenzene, 1-Bromobenzene, Bromobenzol, Monobromobenzene, NSC 6529, Phenyl bromide |
| InChI Formula | InChI=1S/C6H5Br/c7-6-4-2-1-3-5-6/h1-5H |
| SMILES | Brc1ccccc1 |
| IUPAC Name | bromobenzene |
| Molecular Weight (g/mol) | 157.01 |
| Show More |