Learn More
1-(Bromomethyl)pyrene, TRC
Specifications
Specifications
| Chemical Name or Material | 1-(Bromomethyl)pyrene |
| CAS | 2595-90-6 |
| Molecular Formula | C17 H11 Br |
| Quantity | 250 mg |
| Synonym | Pyrene, 1-(bromomethyl)-, 1-(Bromomethyl)pyrene, 1-Pyrenylmethyl bromide, 3-(Bromomethyl)pyrene |
| InChI Formula | InChI=1S/C17H11Br/c18-10-14-7-6-13-5-4-11-2-1-3-12-8-9-15(14)17(13)16(11)12/h1-9H,10H2 |
| SMILES | BrCc1ccc2ccc3cccc4ccc1c2c34 |
| IUPAC Name | 1-(bromomethyl)pyrene |
| Molecular Weight (g/mol) | 295.17 |
| Formula Weight | 294.0044 |