Learn More
1-Bromooctane-3,3,4,4-d4, CDN
Product Code. 30328564
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.1g
0.25g
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30328564
Brand: CDN CDND71330.25G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 1-Bromooctane-3,3,4,4-D4
CAS 1219803-37-8
Molecular Formula C8 D4 H13 Br
Quantity 0.25 g
Synonym 1-Bromooctane-3,3,4,4-d4, 8-Bromooctane-5,5,6,6-d4, 1-Bromooctane-d4, Octyl bromide-d4, 1-Bromooctane-3,3,4,4-D4
InChI Formula InChI=1 S/C8H17Br/c1-2-3-4-5-6-7-8-9/h2-8H2,1H3/i5D2,6D2
SMILES [2 H]C([2 H])(CCBr)C([2 H])([2 H])CCCC
IUPAC Name 1-bromo-3,3,4,4-tetradeuteriooctane
Molecular Weight (g/mol) 197.1492
Formula Weight 196.0765 g/mol
Percent Purity 98 atom % D, min 98% Chemical Purity