missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Cyclohexyl-3-[[4-[2-[(3,5-dichloro-2-methoxybenzoyl)amino]ethyl]phenyl]sulfonyl]urea (1-[[4-[2-[(3,5-Dichloro-2-methoxybenzoyl)amino]ethyl]phenyl]sulfonyl]-3-cyclohexylurea), Mikromol™
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Description
1-Cyclohexyl-3-[[4-[2-[(3,5-dichloro-2-methoxybenzoyl)amino]ethyl]phenyl]sulfonyl]urea (1-[[4-[2-[(3,5-Dichloro-2-methoxybenzoyl)amino]ethyl]phenyl]sulfonyl]-3-cyclohexylurea)
Specifications
Specifications
| Chemical Name or Material | 3-Chloro Glyburide |
| CAS | 57334-89-1 |
| Analyte or Component Names | 3-Chloro Glyburide |
| Shipping Condition | Room Temperature |
| Molecular Formula | C23 H27 Cl2 N3 O5 S |
| Certifications/Compliance | ISO 17025 |
| Synonym | Sabutamol dimer, Salbutamol Imp. F (EP), Glibenclamide Imp. E (EP), 1-[[4-[2-[(3,5-Di-chloro-2-methoxybenzoyl)amino]ethyl]phenyl]sulfonyl]-3-cyclohexylurea, Glibenclamide Imp. E (Pharmeuropa), Glibenclamide Impurity E |
| InChI Formula | InChI=1S/C26H40N2O5/c1-25(2,3)27-13-23(31)17-7-9-21(29)19(11-17)15-33-16-20-12-18(8-10-22(20)30)24(32)14-28-26(4,5)6/h7-12,23-24,27-32H,13-16H2,1-6H3 |
| SMILES | CC(C)(C)NCC(O)c1ccc(O)c(COCc2cc(ccc2O)C(O)CNC(C)(C)C)c1 |
| IUPAC Name | 4-[2-(tert-butylamino)-1-hydroxyethyl]-2-[[5-[2-(tert-butylamino)-1-hydroxyethyl]-2-hydroxyphenyl]methoxymethyl]phenol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction