Learn More
1-Deoxygalactonojirimycin Hydrochloride, TRC
Product Code. 30358381
Change view
Click to view available options
Quantity:
1 mg
10 mg
50 mg
Unit Size:
10mg
1mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30358381 50 mg 50mg
30357817 1 mg 1mg
30350253 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30358381 Supplier Toronto Research Chemicals Supplier No. TRCD23650050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 75172-81-5
Molecular Formula C6 H13 N O4 . Cl H
Molecular Weight (g/mol) 199.63
Synonym 3,4,5-Piperidinetriol, 2-(hydroxymethyl)-, hydrochloride (1:1), (2R,3S,4R,5S)-, 3,4,5-Piperidinetriol, 2-(hydroxymethyl)-, hydrochloride, (2R,3S,4R,5S)- (9CI), 3,4,5-Piperidinetriol, 2-(hydroxymethyl)-, hydrochloride, [2R-(2α,3α,4α,5β)]-, 1-Deoxygalactonojirimycin hydrochloride, Migalastat hydrochloride
IUPAC Name (2R,3S,4R,5S)-2-(hydroxymethyl)piperidine-3,4,5-triol;hydrochloride
SMILES Cl.OC[C@H]1NC[C@H](O)[C@@H](O)[C@H]1O
Specifications
Quantity 50 mg
InChI Formula InChI=1S/C6H13NO4.ClH/c8-2-3-5(10)6(11)4(9)1-7-3;/h3-11H,1-2H2;1H/t3-,4+,5+,6-;/m1./s1
Formula Weight 199.0611
Percent Purity >95%
Chemical Name or Material Deoxygalactonojirimycin hydrochloride
Grade HPLC