missing translation for 'onlineSavingsMsg'
Learn More

1-Ethyl-2-pyrrolidinone, 98%, Thermo Scientific Chemicals

Catalog Number p-3778051
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy
2687-91-4
C6H11NO
113.16
MFCD00003199
ZFPGARUNNKGOBB-UHFFFAOYSA-N
1-ethyl-2-pyrrolidinone, 1-ethyl-2-pyrrolidone, n-ethyl-2-pyrrolidone, n-ethylpyrrolidone, 2-pyrrolidinone, 1-ethyl, n-ethylpyrrolidinone, ethyl pyrrolidone, unii-h0229sx1cw, n-ethyl-, a-pyrrolidone, n-ethyl-2-pyrrolidinone
17595
1-ethylpyrrolidin-2-one
CCN1CCCC1=O
This item is not returnable. View return policy
2687-91-4
C6H11NO
113.16
MFCD00003199
ZFPGARUNNKGOBB-UHFFFAOYSA-N
1-ethyl-2-pyrrolidinone, 1-ethyl-2-pyrrolidone, n-ethyl-2-pyrrolidone, n-ethylpyrrolidone, 2-pyrrolidinone, 1-ethyl, n-ethylpyrrolidinone, ethyl pyrrolidone, unii-h0229sx1cw, n-ethyl-, a-pyrrolidone, n-ethyl-2-pyrrolidinone
17595
1-ethylpyrrolidin-2-one
CCN1CCCC1=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point -75.0°C
Density 0.9980g/mL
Boiling Point 212.5°C
Flash Point 91°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4650 to 1.4670
Quantity 25 g
Fieser 09,220
Specific Gravity 0.998
Solubility Information Solubility in water: soluble. Other solubilities: soluble in organic solvents
Viscosity 2.1 mPa.s (20°C)
Formula Weight 113.16
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1-Ethyl-2-pyrrolidinone
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1-Ethyl-2-pyrrolidinone
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Reproductive toxicity Category 1B
Hazard Statement
  • H360Df-May damage the unborn child. Suspected of damaging fertility.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'