missing translation for 'onlineSavingsMsg'
Learn More

1-Fluoro-2-nitrobenzene, 99%, Thermo Scientific Chemicals

Catalog Number p-3757969
Click to view available options
Quantity:
10 mL
50 mL
250 mL
Packaging:
Glass bottle
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
1493-27-2
C6H4FNO2
141.1
MFCD00007048
PWKNBLFSJAVFAB-UHFFFAOYSA-N
2-fluoronitrobenzene, o-fluoronitrobenzene, o-nitrofluorobenzene, 2-nitrofluorobenzene, benzene, 1-fluoro-2-nitro, benzene, o-nitrofluoro, 2-fluoro-1-nitrobenzene, fluoronitrobenzene, 1-fluoro-2-nitro-benzene, benzene, fluoronitro
73895
1-fluoro-2-nitrobenzene
C1=CC=C(C(=C1)[N+](=O)[O-])F
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
1493-27-2
C6H4FNO2
141.1
MFCD00007048
PWKNBLFSJAVFAB-UHFFFAOYSA-N
2-fluoronitrobenzene, o-fluoronitrobenzene, o-nitrofluorobenzene, 2-nitrofluorobenzene, benzene, 1-fluoro-2-nitro, benzene, o-nitrofluoro, 2-fluoro-1-nitrobenzene, fluoronitrobenzene, 1-fluoro-2-nitro-benzene, benzene, fluoronitro
73895
1-fluoro-2-nitrobenzene
C1=CC=C(C(=C1)[N+](=O)[O-])F

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point -9°C to 6°C
Color Brown to Yellow
Density 1.3350g/mL
Boiling Point 215°C
Flash Point 94°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula FC6H4NO2
Refractive Index 1.5307 to 1.5327
Quantity 10 mL
Beilstein 05, 241
Specific Gravity 1.335
Solubility Information Solubility in water: immiscible
Formula Weight 141.1
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 1-Fluoro-2-nitrobenzene
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'