Learn More
1-Heptadecyl Bromide, TRC
Product Code. 30407260
Change view
Click to view available options
Quantity:
2 g
Unit Size:
2g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30407260 2 g 2g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30407260 Supplier Toronto Research Chemicals Supplier No. TRCH2790002G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 1-Heptadecyl Bromide
CAS 3508-00-7
Molecular Formula C17 H35 Br
Quantity 2 g
InChI Formula InChI=1S/C17H35Br/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18/h2-17H2,1H3
SMILES CCCCCCCCCCCCCCCCCBr
IUPAC Name 1-bromoheptadecane
Molecular Weight (g/mol) 319.36
Formula Weight 318.1922