Learn More
1-Iodoheptafluoropropane, TRC
Specifications
Specifications
| Chemical Name or Material | 1-Iodoheptafluoropropane |
| CAS | 754-34-7 |
| Molecular Formula | C3 F7 I |
| Quantity | 1 g |
| Synonym | Perfluoro-1-iodopropane, Heptafluoro-1-iodopropane, Perfluoropropyl Iodide |
| InChI Formula | InChI=1S/C3F7I/c4-1(5,2(6,7)8)3(9,10)11 |
| SMILES | FC(F)(F)C(F)(F)C(F)(F)I |
| IUPAC Name | 1,1,1,2,2,3,3-heptafluoro-3-iodopropane |
| Molecular Weight (g/mol) | 295.925 |
| Formula Weight | 295.893 |