Learn More
1-Iodopropane-2,2-d2, CDN
Specifications
Specifications
| Chemical Name or Material | 1-Iodopropane-2,2-D2 |
| CAS | 25493-15-6 |
| Molecular Formula | C3 D2 H5 I |
| Quantity | 1 g |
| Synonym | Propane-2,2-d2, 1-iodo- (8 CI, 9 CI), 1-Iodopropane-2,2-d2, 1-Iodopropane-d2, 1-Iodopropane-2,2-D2, [2,2-2H2]Propyl iodide |
| InChI Formula | InChI=1 S/C3H7I/c1-2-3-4/h2-3H2,1H3/i2D2 |
| SMILES | [2 H]C([2 H])(C)CI |
| IUPAC Name | 2,2-dideuterio-1-iodopropane |
| Molecular Weight (g/mol) | 172.0045 |
| Formula Weight | 171.9718 g/mol |
| Show More |