Learn More
1-Iodopropane-3,3,3-d3, CDN
Specifications
Specifications
| Chemical Name or Material | 1-Iodopropane-3,3,3-D3 |
| CAS | 25493-16-7 |
| Molecular Formula | C3 D3 H4 I |
| Quantity | 1 g |
| Synonym | Propane-1,1,1-d3, 3-iodo- (6 CI, 8 CI, 9 CI), 1-Iodopropane-3,3,3-d3, 1-Iodopropane-d3, 1-Iodopropane-3,3,3-D3, [3,3,3-2H3]Propyl iodide |
| InChI Formula | InChI=1 S/C3H7I/c1-2-3-4/h2-3H2,1H3/i1D3 |
| SMILES | [2 H]C([2 H])([2 H])CCI |
| IUPAC Name | 1,1,1-trideuterio-3-iodopropane |
| Molecular Weight (g/mol) | 173.011 |
| Formula Weight | 172.978 g/mol |
| Show More |