Learn More
1-(Isocyanomethyl)-1H-benzotriazole, TRC
Specifications
Specifications
| Chemical Name or Material | 1-(Isocyanomethyl)-1H-benzotriazole |
| CAS | 87022-42-2 |
| Molecular Formula | C8H6N4 |
| Quantity | 100 mg |
| Synonym | 1H-Benzotriazol-1-ylmethyl Isocyanide, Benzotriazol-1-ylmethyl Isocyanide |
| InChI Formula | InChI=1S/C8H6N4/c1-9-6-12-8-5-3-2-4-7(8)10-11-12/h2-5H,6H2 |
| SMILES | [C-]#[N+]Cn1nnc2ccccc12 |
| IUPAC Name | 1-(isocyanomethyl)benzotriazole |
| Molecular Weight (g/mol) | 158.16 |
| Formula Weight | 158.0592 |