Learn More
((1-Methoxy-2-methylprop-1-en-1-yl)oxy)trimethylsilane, 97%, BLD Pharm™
Specifications
Specifications
| CAS | 31469-15-5 |
| Molecular Formula | C8H18O2Si |
| MDL Number | MFCD00010232 |
| Quantity | 5 g |
| SMILES | C[Si](C)(OC(OC)=C(C)C)C |
| Molecular Weight (g/mol) | 174.31 |
| Percent Purity | 97% |
| Chemical Name or Material | ((1-Methoxy-2-methylprop-1-en-1-yl)oxy)trimethylsilane |