missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Methyl-1-propenylmagnesium bromide, 0.5M solution in THF, AcroSeal™, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass Bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
CAS Min % | 7.68, 90.62 |
CAS Max % | 9.38, 92.32 |
Color | Colorless to Yellow |
Density | 0.9340g/mL |
Boiling Point | 67.0°C |
Flash Point | −26°C |
Packaging | AcroSeal™ Glass Bottle |
Quantity | 800 mL |
Specific Gravity | 0.934 |
Solubility Information | Solubility in water: reacts |
Show More |
Safety and Handling
Product Identifier
- 1-Methyl-1-propenylmagnesium bromide, 0.5M solution in THF
- Danger
- Acute toxicity Category 4
- Carcinogenicity Category 2
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 2
- Skin corrosion/irritation Category 1 B
- Specific target organ toxicity Category 3
- Substances or mixtures which\, in contact with water\, emits flammable gases Category 3
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H351-Suspected of causing cancer.
- H335-May cause respiratory irritation.
- H336-May cause drowsiness or dizziness.
- H302-Harmful if swallowed.
- H261-In contact with water releases flammable gases.
- H225-Highly flammable liquid and vapour.
- EUH014-Reacts violently with water.
- EUH019-May form explosive peroxides.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P231+P232-Handle under inert gas. Protect from moisture.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P402+P404-Store in a dry place. Store in a closed container.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'