missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-1H-imidazole-4-sulfonyl chloride, 95+%, Thermo Scientific™

Catalog Number p-4509857
Click to view available options
Quantity:
250 mg
1 g
10 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
137049-00-4
C4H5ClN2O2S
180.61
MFCD00068060
KXUGUWTUFUWYRS-UHFFFAOYSA-N
1-methyl-1h-imidazole-4-sulfonyl chloride, 1-methyl-1h-imidazole-4-sulphonyl chloride, 1h-imidazole-4-sulfonyl chloride, 1-methyl, 1-methylimidazole-4-sulfonylchloride, 1-methylimidazole-4-sulphonyl chloride, chloro 1-methylimidazol-4-yl sulfone, 2-methoxymethylpropenal, acmc-209cak, buttpark 27\06-11, 1-methylimidazol-4-sulfonylchloride
2736887
1-methylimidazole-4-sulfonyl chloride
CN1C=NC(=C1)S(Cl)(=O)=O
This item is not returnable. View return policy
137049-00-4
C4H5ClN2O2S
180.61
MFCD00068060
KXUGUWTUFUWYRS-UHFFFAOYSA-N
1-methyl-1h-imidazole-4-sulfonyl chloride, 1-methyl-1h-imidazole-4-sulphonyl chloride, 1h-imidazole-4-sulfonyl chloride, 1-methyl, 1-methylimidazole-4-sulfonylchloride, 1-methylimidazole-4-sulphonyl chloride, chloro 1-methylimidazol-4-yl sulfone, 2-methoxymethylpropenal, acmc-209cak, buttpark 27\06-11, 1-methylimidazol-4-sulfonylchloride
2736887
1-methylimidazole-4-sulfonyl chloride
CN1C=NC(=C1)S(Cl)(=O)=O
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 180.61
Percent Purity 95%
Chemical Name or Material 1-Methyl-1H-imidazole-4-sulfonyl chloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'