missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-1H-indazole-4-boronic acid pinacol ester, 97%, Thermo Scientific™

Catalog Number p-4812619
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
885698-94-2
C14H19BN2O2
258.13
MFCD10700158
OZHFBNULPUBRJF-UHFFFAOYSA-N
1-methyl-1h-indazole-4-boronic acid pinacol ester, 1-methyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-indazole, 1-methylindazole-4-boronic acid pinacol ester, 1-methyl-4-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-indazole, 1-methyl-1h-indazole-4-boronic acid, pinacol ester, 1-methyl-4-tetramethyl-1,3,2-dioxaborolan-2-yl indazole, 1-methyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl indazole, 1-methyl-4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-1h-indazole, 1h-indazole, 1-methyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl
45480187
1-methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)indazole
CN1N=CC2=C(C=CC=C12)B1OC(C)(C)C(C)(C)O1
This item is not returnable. View return policy
885698-94-2
C14H19BN2O2
258.13
MFCD10700158
OZHFBNULPUBRJF-UHFFFAOYSA-N
1-methyl-1h-indazole-4-boronic acid pinacol ester, 1-methyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-indazole, 1-methylindazole-4-boronic acid pinacol ester, 1-methyl-4-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-indazole, 1-methyl-1h-indazole-4-boronic acid, pinacol ester, 1-methyl-4-tetramethyl-1,3,2-dioxaborolan-2-yl indazole, 1-methyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl indazole, 1-methyl-4-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-1h-indazole, 1h-indazole, 1-methyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl
45480187
1-methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)indazole
CN1N=CC2=C(C=CC=C12)B1OC(C)(C)C(C)(C)O1
Melting Point 83.0°C to 86.0°C
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 258.13
Percent Purity 97%
Chemical Name or Material 1-Methyl-1H-indazole-4-boronic acid pinacol ester
compliance-icons
Product Identifier
  • 1-Methyl-1H-indazole-4-boronic acid pinacol ester
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'