Learn More
1-Methyl-5-amino-1H-benzimidazole-2-butanoic Acid Ethyl Ester, Mikromol™
Product Code. 30460813
Change view
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30460813 4 x 25 mg -
30051555 25 mg 25mg
30049788 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30460813 Supplier Mikromol Supplier No. MM0154.0300254AMP
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
1-Methyl-5-amino-1H-benzimidazole-2-butanoic Acid Ethyl Ester, Mikromol
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 1-Methyl-5-amino-1H-benzimidazole-2-butanoic Acid Ethyl Ester
CAS 3543-73-5
Analyte or Component Names 1-Methyl-5-amino-1H-benzimidazole-2-butanoic Acid Ethyl Ester
Shipping Condition Room Temperature
Molecular Formula C14 H19 N3 O2
Certifications/Compliance ISO 17025
Synonym 1-Methyl-5-amino-1H-benzimidazole-2-butanoic Acid Ethyl Ester, 2-Benzimidazolebutyric acid, 5-amino-1-methyl-, ethyl ester (7CI,8CI), 4-[5-Amino-1-methylbenzimidazol-2-yl]butanoic acid ethyl ester, Ethyl 1-methyl-5-amino-1H-benzimidazole-2-butanoate
InChI Formula InChI=1S/C14H19N3O2/c1-3-19-14(18)6-4-5-13-16-11-9-10(15)7-8-12(11)17(13)2/h7-9H,3-6,15H2,1-2H3
Solution Type Neat
SMILES CCOC(=O)CCCc1nc2cc(N)ccc2n1C
IUPAC Name ethyl 4-(5-amino-1-methylbenzimidazol-2-yl)butanoate
Molecular Weight (g/mol) 261.32
Quantity 4 x 25 mg
Formula Weight 261.1477 g/mol