Learn More
1-Methyl-5-amino-1H-benzimidazole-2-butanoic Acid Ethyl Ester, TRC
Specifications
Specifications
| Chemical Name or Material | 1-Methyl-5-amino-1H-benzimidazole-2-butanoic Acid Ethyl Ester |
| CAS | 3543-73-5 |
| Molecular Formula | C14 H19 N3 O2 |
| Quantity | 5 mg |
| Synonym | 1-Methyl-5-amino-1H-benzimidazole-2-butanoic Acid Ethyl Ester, 2-Benzimidazolebutyric acid','5-amino-1-methyl-','ethyl ester (7CI,8CI), 4-[5-Amino-1-methylbenzimidazol-2-yl]butanoic acid ethyl ester, Ethyl 1-methyl-5-amino-1H-benzimidazole-2-butanoate |
| InChI Formula | InChI=1S/C14H19N3O2/c1-3-19-14(18)6-4-5-13-16-11-9-10(15)7-8-12(11)17(13)2/h7-9H,3-6,15H2,1-2H3 |
| SMILES | CCOC(=O)CCCc1nc2cc(N)ccc2n1C |
| IUPAC Name | ethyl 4-(5-amino-1-methylbenzimidazol-2-yl)butanoate |
| Molecular Weight (g/mol) | 261.32 |
| Formula Weight | 261.1477 |