missing translation for 'onlineSavingsMsg'
Learn More

1-Methylindole-3-acetic Acid, TRC

Product Code. 30466629
Click to view available options
Quantity:
1 g
Unit Size:
1g
This item is not returnable. View return policy

Product Code. 30466629

Brand: Toronto Research Chemicals TRCM3134951G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1-Methylindole-3-acetic Acid
CAS 1912-48-7
Molecular Formula C11H11NO2
Quantity 1 g
Synonym (1-Methyl-1H-indol-3-yl)acetic Acid, (1-Methylindol-4-yl)acetic Acid, 1-Methyl-1H-indole-3-acetic Acid, 1-Methylindole-3-acetic Acid, 1-Methylindolyl-3-acetic Acid, 2-(1-Methyl-1H-indol-3-yl)acetic Acid, 2-(1-Methyl-3-indolyl)acetic Acid, 2-(N-Methylindole-3-yl)acetic Acid, N-Methylindole-3-acetic Acid, N-Methylindoleacetic Acid, NSC 78007
InChI Formula InChI=1S/C11H11NO2/c1-12-7-8(6-11(13)14)9-4-2-3-5-10(9)12/h2-5,7H,6H2,1H3,(H,13,14)
SMILES Cn1cc(CC(=O)O)c2ccccc12
IUPAC Name 2-(1-methylindol-3-yl)acetic acid
Molecular Weight (g/mol) 189.21
Formula Weight 189.079
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.