Learn More
1-n-Butylpyridinium bromide, 99%
Product Code. 11401640
Change view
Click to view available options
Quantity:
5 g
50 g
Unit Size:
50g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11401640 5 g 5g
11411640 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11401640 Supplier Thermo Scientific Alfa Aesar Supplier No. H26995.06
Request Bulk or Custom Format
CAS: 874-80-6 | C9H14BrN | 216.12 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 874-80-6
Molecular Formula C9H14BrN
Molecular Weight (g/mol) 216.12
MDL Number MFCD00671496
InChI Key KVBQNFMTEUEOCD-UHFFFAOYSA-M
Synonym 1-butylpyridinium bromide, butylpyridinium bromide, 1-butylpyridin-1-ium bromide, n-butylpyridinium bromide, unii-f72fd1bswg, f72fd1bswg, 1-n-butylpyridinium bromide, pyridinium, 1-butyl-, bromide, 1-butylpyridiniumbromide, acmc-209qnu
PubChem CID 2734238
IUPAC Name 1-butylpyridin-1-ium;bromide
SMILES [Br-].CCCC[N+]1=CC=CC=C1
Specifications
Melting Point 105°C to 108°C
Quantity 5 g
Sensitivity Hygroscopic
Formula Weight 216.12
Percent Purity 99%
Chemical Name or Material 1-n-Butylpyridinium bromide