Learn More
1-n-Hexyl-3-methylimidazolium tetrafluoroborate, 99%
Product Code. 11461840
Change view
Click to view available options
Quantity:
5 g
50 g
Unit Size:
50g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11461840 5 g 5g
11471840 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11461840 Supplier Thermo Scientific Alfa Aesar Supplier No. H27548.06
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
CAS: 244193-50-8 | C10H19BF4N2 | 254.08 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 244193-50-8
Molecular Formula C10H19BF4N2
Molecular Weight (g/mol) 254.08
MDL Number MFCD03095459
InChI Key MFXLOVLEQJRXFP-UHFFFAOYSA-N
Synonym 1-hexyl-3-methylimidazolium tetrafluoroborate, 1-n-hexyl-3-methylimidazolium tetrafluoroborate, hmimbf4, c6mim bf4, acmc-1ckkl, hmim bf4, hmim+bf4, dsstox_cid_29108, dsstox_rid_83327, dsstox_gsid_49252
PubChem CID 2734179
IUPAC Name 1-hexyl-3-methylimidazol-3-ium;tetrafluoroborate
SMILES F[B-](F)(F)F.CCCCCCN1C=C[N+](C)=C1
Specifications
Melting Point -81°C
Density 1.149
Refractive Index 1.43
Quantity 5 g
Formula Weight 254.08
Percent Purity 99%
Chemical Name or Material 1-n-Hexyl-3-methylimidazolium tetrafluoroborate