Learn More
1-Nitronaphthalen-2-ol, 98%, BLD Pharm™
Specifications
Specifications
| CAS | 550-60-7 |
| Molecular Formula | C10H7NO3 |
| MDL Number | MFCD00021461 |
| Quantity | 250 mg |
| SMILES | OC1=CC=C2C=CC=CC2=C1[N+]([O-])=O |
| Molecular Weight (g/mol) | 189.17 |
| Percent Purity | 98% |
| Chemical Name or Material | 1-Nitronaphthalen-2-ol |