Learn More
1-Nonanol, 97%
Product Code. p-3758575 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
250 mL
1 L
Packaging:
Glass bottle
Unit Size:
1 L
100mL
250mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10498690 100 mL Glass bottle 100mL
10744471 250 mL Glass bottle 250mL
10541554 1 L Glass bottle 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10498690 Supplier Thermo Scientific Acros Supplier No. 157471000
Add to basket
CAS: 143-08-8 | C9H20O | 144.26 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 143-08-8
Molecular Formula C9H20O
Molecular Weight (g/mol) 144.26
MDL Number MFCD00002990
InChI Key ZWRUINPWMLAQRD-UHFFFAOYSA-N
Synonym 1-nonanol, nonanol, n-nonyl alcohol, nonyl alcohol, pelargonic alcohol, octyl carbinol, 1-hydroxynonane, nonalol, n-nonanol, n-nonan-1-ol
PubChem CID 8914
ChEBI CHEBI:35986
IUPAC Name nonan-1-ol
SMILES CCCCCCCCCO
Specifications
CAS Min % 96.0
CAS Max % 100.0
Melting Point -5.0°C
Color Colorless
Density 0.8270g/mL
Boiling Point 213.0°C to 215.0°C
Flash Point 74°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula CH3(CH2)8OH
Refractive Index 1.4324 to 1.4344
Quantity 100 mL
UN Number 3082
Beilstein 01, 423
Merck Index 15, 6766
Specific Gravity 0.827
Solubility Information Solubility in water: 1g/L (20°C)
Formula Weight 144.26
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 1-Nonanol
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1-Nonanol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H411-Toxic to aquatic life with long lasting effects.
  • H332-Harmful if inhaled.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.