Learn More
1-O-Acetyl-2,3,5-tri-O-benzoyl-L-ribose, TRC
Specifications
Specifications
| Chemical Name or Material | 1-O-Acetyl-2,3,5-tri-O-benzoyl-L-ribose |
| CAS | 26287-72-9 |
| Molecular Formula | C28H24O9 |
| Quantity | 1 g |
| InChI Formula | InChI=1S/C28H24O9/c1-18(29)34-28-24(37-27(32)21-15-9-4-10-16-21)23(36-26(31)20-13-7-3-8-14-20)22(35-28)17-33-25(30)19-11-5-2-6-12-19/h2-16,22-24,28H,17H2,1H3/t22-,23-,24-,28?/m0/s1 |
| SMILES | CC(=O)OC1O[C@@H](COC(=O)c2ccccc2)[C@H](OC(=O)c3ccccc3)[C@@H]1OC(=O)c4ccccc4 |
| IUPAC Name | [(2S,3S,4S)-5-acetyloxy-3,4-dibenzoyloxyoxolan-2-yl]methyl benzoate |
| Molecular Weight (g/mol) | 504.48 |
| Formula Weight | 504.142 |