Learn More
1-O-Acetyl-2,3,5-tri-O-benzoyl-L-ribose, TRC
Product Code. 30323182
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30323182 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30323182 Supplier Toronto Research Chemicals Supplier No. TRCA188645250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 1-O-Acetyl-2,3,5-tri-O-benzoyl-L-ribose
CAS 26287-72-9
Molecular Formula C28H24O9
Quantity 250 mg
InChI Formula InChI=1S/C28H24O9/c1-18(29)34-28-24(37-27(32)21-15-9-4-10-16-21)23(36-26(31)20-13-7-3-8-14-20)22(35-28)17-33-25(30)19-11-5-2-6-12-19/h2-16,22-24,28H,17H2,1H3/t22-,23-,24-,28?/m0/s1
SMILES CC(=O)OC1O[C@@H](COC(=O)c2ccccc2)[C@H](OC(=O)c3ccccc3)[C@@H]1OC(=O)c4ccccc4
IUPAC Name [(2S,3S,4S)-5-acetyloxy-3,4-dibenzoyloxyoxolan-2-yl]methyl benzoate
Molecular Weight (g/mol) 504.48
Formula Weight 504.142