missing translation for 'onlineSavingsMsg'
Learn More

1-Octanesulfonic acid, sodium salt, HPLC grade, Thermo Scientific Chemicals

Catalog Number p-3211410
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
5324-84-5
C8H17NaO3S
216.27
MFCD00007544
HRQDCDQDOPSGBR-UHFFFAOYSA-M
sodium 1-octanesulfonate, sodium octane-1-sulfonate, 1-octanesulfonic acid sodium salt, sodium octanesulfonate, 1-octanesulfonic acid, sodium salt, 1-octanesulfonate, sodium, sodium octylsulfonate, unii-j46ez5jyb3, octyl sodium sulfonate, ipc-alks-8
23669624
[Na+].CCCCCCCCS([O-])(=O)=O
5324-84-5
C8H17NaO3S
216.27
MFCD00007544
HRQDCDQDOPSGBR-UHFFFAOYSA-M
sodium 1-octanesulfonate, sodium octane-1-sulfonate, 1-octanesulfonic acid sodium salt, sodium octanesulfonate, 1-octanesulfonic acid, sodium salt, 1-octanesulfonate, sodium, sodium octylsulfonate, unii-j46ez5jyb3, octyl sodium sulfonate, ipc-alks-8
23669624
[Na+].CCCCCCCCS([O-])(=O)=O

1-Octanesulfonic acid, sodium salt, HPLC grade, Acros Organics, Quantity: 25g, White, Melting Point: >300.0 deg.C, Molecular Weight: 216.28, Percent Purity: >-98.0%

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For ion pair chromatography
TRUSTED_SUSTAINABILITY
Absorbance (0.25M aqueous solution), (0.25 M aqueous solution) at 0nm, 0.04 max. at 260nm, 0.05 max. at 240nm, 0.05 max. at 250nm, 0.075 max. at 230nm, 0.1 max. at 220nm, 0.2 max. at 210nm, 0.3 max. at 200nm
Melting Point >300.0°C
Color White
Infrared Spectrum Authentic
Packaging Glass bottle
Linear Formula CH3(CH2)7SO3Na
Quantity 5 g
Solubility Information Solubility in water: freely soluble. Other solubilities: 4g/L in methanol
IUPAC Name sodium;octane-1-sulfonate
Formula Weight 216.28
Percent Purity ≥98.0%
Physical Form Crystalline Powder
Chemical Name or Material 1-Octanesulfonic acid, sodium salt
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'