missing translation for 'onlineSavingsMsg'
Learn More

1-Octanol, ≥99%, ACS Spectrophotometric Grade, Honeywell Riedel-de Haën™

Product Code. p-7120034
Click to view available options
Quantity:
1 L
2 L
Packaging:
Glass bottle
Unit Size:
1 L
2 L
This item is not returnable. View return policy

Product Code. 15665090

Brand: Honeywell Riedel-de Haën 3605621L

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

ACS Spectrophotometric Grade,≥99%

  • ACS grade quality reagents and solvents tested according to the specific use for spectrophotometric applications.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111-87-5
Molecular Formula C8H18O
Molecular Weight (g/mol) 130.23
MDL Number MFCD00002988
InChI Key KBPLFHHGFOOTCA-UHFFFAOYSA-N
Synonym 1-octanol, octanol, octyl alcohol, n-octanol, capryl alcohol, caprylic alcohol, n-octyl alcohol, heptyl carbinol, 1-hydroxyoctane, primary octyl alcohol
PubChem CID 957
ChEBI CHEBI:16188
IUPAC Name octan-1-ol
SMILES CCCCCCCCO

Specifications

Melting Point -15°C
Density 0.827g/mL at 25°C
Boiling Point 194°C
Flash Point 80°C (176°F)
Packaging Glass bottle
Linear Formula CH3(CH2)7OH
Refractive Index 1.429
Quantity 1 L
Beilstein 1697461
Vapor Density 4.5 (vs air)
Formula Weight 130.23g/mol
Percent Purity ≥99%
Grade ACS
Chemical Name or Material 1-Octanol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.