Learn More
1-(p-Toluenesulfonyl)pyrrole, 98%
Product Code. 11412100
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11412100 5 g 5g
11422100 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11412100 Supplier Thermo Scientific Alfa Aesar Supplier No. H28641.06
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
CAS: 17639-64-4 | C11H11NO2S | 221.274 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 17639-64-4
Molecular Formula C11H11NO2S
Molecular Weight (g/mol) 221.274
MDL Number MFCD00145014
InChI Key OXWIEWFMRVJGNY-UHFFFAOYSA-N
Synonym 1-tosylpyrrole, 1-tosyl-1h-pyrrole, 1-p-toluenesulfonyl pyrrole, n-tosylpyrrole, 1-4-methylphenyl sulfonyl-1h-pyrrole, 1-p-tolylsulfonyl pyrrole, 1-tosyl-pyrrole, 1-4-methylphenylsulfonyl pyrrole, 1-4-methylbenzenesulfonyl pyrrole, 1h-pyrrole, 1-4-methylphenyl sulfonyl
PubChem CID 640087
IUPAC Name 1-(4-methylphenyl)sulfonylpyrrole
SMILES CC1=CC=C(C=C1)S(=O)(=O)N2C=CC=C2
Specifications
Melting Point 99°C to 103°C
Quantity 5 g
Formula Weight 221.28
Percent Purity 98%
Chemical Name or Material 1-(p-Toluenesulfonyl)pyrrole