missing translation for 'onlineSavingsMsg'
Learn More

1-Palmitoyl-2-oleoyl-sn-glycero-3-phospho-(1'-rac-glycerol) Sodium Salt, TRC

Product Code. 30424167
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30424167 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30424167 Supplier Toronto Research Chemicals Supplier No. TRCP16098025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1-Palmitoyl-2-oleoyl-sn-glycero-3-phospho-(1'-rac-glycerol) Sodium Salt
CAS 268550-95-4
Molecular Formula C40H76NaO10P
Quantity 25 mg
Synonym (9Z)-9-Octadecenoic acid (1R)-1-[[[(2,3-dihydroxypropoxy)hydroxyphosphinyl]oxy]methyl]-2-[(1-oxohexadecyl)oxy]ethyl ester sodium salt, (9Z)-(9-Octadecenoic acid 1R)-1-[[[(2,3-dihydroxypropoxy)hydroxyphosphinyl]oxy]methyl]-2-[(1-oxohexadecyl)oxy]ethyl ester monosodium salt
InChI Formula InChI=1S/C40H77O10P.Na/c1-3-5-7-9-11-13-15-17-18-20-22-24-26-28-30-32-40(44)50-38(36-49-51(45,46)48-34-37(42)33-41)35-47-39(43)31-29-27-25-23-21-19-16-14-12-10-8-6-4-2;/h17-18,37-38,41-42H,3-16,19-36H2,1-2H3,(H,45,46);/q;+1/p-1/b18-17-;/t37?,38-;/m1./s1
SMILES COC(OC)[C@@H](C)[C@@H]1[C@@]2(C)[C@@H](CC1)C(=O)CCC2
IUPAC Name sodium;2,3-dihydroxypropyl [(2R)-3-hexadecanoyloxy-2-[(Z)-octadec-9-enoyl]oxypropyl] phosphate
Molecular Weight (g/mol) 770.99
Formula Weight 770.5074
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.