Learn More
1-Piperidinepropanol, TRC
Specifications
Specifications
| Chemical Name or Material | 1-Piperidinepropanol |
| CAS | 104-58-5 |
| Molecular Formula | C8 H17 N O |
| Quantity | 5 g |
| Synonym | 1-(3-Hydroxypropyl)piperidine, 3-(1-Piperidinyl)propanol, 3-(1-Piperidyl)propan-1-ol, 3-(Piperidin-1-yl)propan-1-ol, 3-Piperidino-1-propanol, 3-Piperidinopropanol, N-(3-Hydroxypropyl)piperidine, NSC 66531 |
| InChI Formula | InChI=1S/C8H17NO/c10-8-4-7-9-5-2-1-3-6-9/h10H,1-8H2 |
| SMILES | OCCCN1CCCCC1 |
| IUPAC Name | 3-piperidin-1-ylpropan-1-ol |
| Molecular Weight (g/mol) | 143.227 |
| Formula Weight | 143.131 |