Learn More
1-Tetracosene, TRC
Product Code. 30456246
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30456246 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30456246 Supplier Toronto Research Chemicals Supplier No. TRCT58576350MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 1-Tetracosene
CAS 10192-32-2
Molecular Formula C24 H48
Quantity 50 mg
InChI Formula InChI=1S/C24H48/c1-3-5-7-9-11-13-15-17-19-21-23-24-22-20-18-16-14-12-10-8-6-4-2/h3H,1,4-24H2,2H3
SMILES CCCCCCCCCCCCCCCCCCCCCCC=C
IUPAC Name tetracos-1-ene
Molecular Weight (g/mol) 336.638
Formula Weight 336.376