Learn More
1-(trans-2-Phenylethenyl)-4-(2-phenylethyl)benzene, 97%
Product Code. 15477785
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15477785 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15477785 Supplier Thermo Scientific Alfa Aesar Supplier No. H31077.03
Request Bulk or Custom Format
CAS: 95166-77-1 | C22H20 | 284.402 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifications
CAS 95166-77-1
Melting Point 155°C to 157°C
Molecular Formula C22H20
MDL Number MFCD12407080
Quantity 1 g
Synonym 1-trans-2-phenylethenyl-4-2-phenylethyl benzene, 1-e-2-phenylethenyl-4-2-phenylethyl benzene, 4-phenethyl-cis-stilbene, 4-phenethyl-trans-stilbene, 4-trans-beta-styryl bibenzyl
InChI Key VKIHNQGTZXZCPE-ACCUITESSA-N
SMILES C1=CC=C(C=C1)CCC2=CC=C(C=C2)C=CC3=CC=CC=C3
IUPAC Name 1-[(E)-2-phenylethenyl]-4-(2-phenylethyl)benzene
Molecular Weight (g/mol) 284.402
PubChem CID 59832893
Formula Weight 284.4
Percent Purity 97%
Chemical Name or Material 1-(trans-2-Phenylethenyl)-4-(2-phenylethyl)benzene