missing translation for 'onlineSavingsMsg'
Learn More

1-(Trifluoromethyl)-1-cyclobutanecarboxylic acid, 95%, Thermo Scientific™

Product Code. 11405590
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11405590 100 mg 100mg
11415590 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11405590 Supplier Thermo Scientific Alfa Aesar Supplier No. H25850.MC
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 277756-45-3
Molecular Formula C6H7F3O2
Molecular Weight (g/mol) 168.115
MDL Number MFCD03425197
InChI Key HYRCPSRIIWIESW-UHFFFAOYSA-N
Synonym 1-trifluoromethyl cyclobutanecarboxylic acid, 1-trifluoromethyl cyclobutane-1-carboxylic acid, 1-trifluoromethyl-1-cyclobutanecarboxylic acid, cyclobutanecarboxylicacid, 1-trifluoromethyl, 1-trifluoromethyl-cyclobutanecarboxylic acid, 1-trifluoromethyl-1-cyclobutane carboxylic acid, 1-trifluoromethyl-1-cyclobutane-1-carboxylic acid, 1-trifluormethyl-cyclobutanecarboxylic acid, 1-trifluoromethylcyclobutyl carboxylic acid, 1-trifluoromethyl-1-cyclobutylcarboxylic acid
PubChem CID 2782838
IUPAC Name 1-(trifluoromethyl)cyclobutane-1-carboxylic acid
SMILES C1CC(C1)(C(=O)O)C(F)(F)F

Specifications

Melting Point 24°C to 26°C
Density 1.33
Boiling Point 76°C (14 mmHg)
Flash Point 96°C (205°F)
Refractive Index 1.391
Quantity 100 mg
Formula Weight 168.12
Percent Purity 95%
Chemical Name or Material 1-(Trifluoromethyl)-1-cyclobutanecarboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.