Learn More
1-Tritylimidazole-4-carboxaldehyde, 98%, Thermo Scientific Chemicals
Catalog Number p-7028131
Click to view available options
Quantity:
1 g
5 g
25 g
33016-47-6
C23H18N2O
338.41
MFCD02179554
YQYLLBSWWRWWAY-UHFFFAOYSA-N
1-tritylimidazole-4-carboxaldehyde, 1-trityl-1h-imidazole-4-carbaldehyde, 1-trityl-1h-imidazole-4-carboxaldehyde, 1-trityl-imdazole-4-carboxadehyde, 4-formyl-1-tritylimidazole, 4-formyl-1-trityl-1h-imidazole, 1-trityl-4-formylimidazole, 1-triphenylmethyl imidazole-4-carbaldehyde, 1-triphenylmethyl-1h-imidazole-4-carbaldehyde, 1h-imidazole-4-carboxaldehyde, 1-triphenylmethyl
618233
O=CC1=CN(C=N1)C(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
33016-47-6
C23H18N2O
338.41
MFCD02179554
YQYLLBSWWRWWAY-UHFFFAOYSA-N
1-tritylimidazole-4-carboxaldehyde, 1-trityl-1h-imidazole-4-carbaldehyde, 1-trityl-1h-imidazole-4-carboxaldehyde, 1-trityl-imdazole-4-carboxadehyde, 4-formyl-1-tritylimidazole, 4-formyl-1-trityl-1h-imidazole, 1-trityl-4-formylimidazole, 1-triphenylmethyl imidazole-4-carbaldehyde, 1-triphenylmethyl-1h-imidazole-4-carbaldehyde, 1h-imidazole-4-carboxaldehyde, 1-triphenylmethyl
618233
O=CC1=CN(C=N1)C(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
1-Tritylimidazole-4-carboxaldehyde is used in the synthesis of histamine and girolline
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1-Tritylimidazole-4-carboxaldehyde is used in the synthesis of histamine and girolline
Notes
Air sensitive. Store under inert gas. Incompatible with oxidizing agents, air.
TRUSTED_SUSTAINABILITY
Melting Point ∼192°C (decomposition)
Quantity 1 g
Sensitivity Air Sensitive
IUPAC Name 1-tritylimidazole-4-carbaldehyde
Formula Weight 338.41
Percent Purity 98%
Chemical Name or Material 1-Tritylimidazole-4-carboxaldehyde