Learn More
1-Vinylnaphthalene, 95%, stab. with 4-tert-butylcatechol
Product Code. p-7033865
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. 15469125
Brand: Thermo Scientific Alfa Aesar H51043.03
Request Bulk or Custom Format
CAS: 826-74-4 | C12H10 | 154.21 g/mol
1-Vinylnaphthalene is used as a precursor of polynuclear aromatic hydrocarbons in tobacco smoke. It is also used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1-Vinylnaphthalene is used as a precursor of polynuclear aromatic hydrocarbons in tobacco smoke. It is also used as pharmaceutical intermediate.
Solubility
Soluble in ethyl acetate, hexane and methanol.
Notes
Moisture sensitive. Incompatible with oxidizing agents, heat, light, ultraviolet radiation and free radical initiators.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 826-74-4
Molecular Formula C12H10
Molecular Weight (g/mol) 154.21
MDL Number MFCD00075766
InChI Key IGGDKDTUCAWDAN-UHFFFAOYSA-N
Synonym 1-vinylnaphthalene, vinylnaphthalene, 1-vinyl naphthalene, unii-pf20cj2k0n, pf20cj2k0n, 1-vinylphthalene, 1-vinyl-naphthalene, naphthalene, ethenyl, naphthalen-1-yl ethene, naphthalene, 1-ethenyl
PubChem CID 70004
ChEBI CHEBI:51327
IUPAC Name 1-ethenylnaphthalene
SMILES C=CC1=C2C=CC=CC2=CC=C1
Specifications
Density 1.04
Boiling Point 135°C to 138°C
Quantity 1 g
Sensitivity Moisture sensitive
Solubility Information Soluble in ethyl acetate,hexane and methanol.
Formula Weight 154.21
Percent Purity 95%
Chemical Name or Material 1-Vinylnaphthalene, stabilized with 4-tert-butylcatechol