Learn More
10,11-Dihydro-10-hydroxy Carbamazepine-D4 (Major), TRC
Specifications
Specifications
| Chemical Name or Material | 10,11-Dihydro-10-hydroxycarbazepine-D4 (Major) |
| CAS | 1020719-39-4 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C15 2H4 H10 N2 O2 |
| Quantity | 5 mg |
| Synonym | Licarbazepine D4 |
| InChI Formula | InChI=1S/C15H14N2O2/c16-15(19)17-12-7-3-1-5-10(12)9-14(18)11-6-2-4-8-13(11)17/h1-8,14,18H,9H2,(H2,16,19)/i1D,3D,5D,7D |
| SMILES | [2H]c1c([2H])c([2H])c2N(C(=O)N)c3ccccc3C(O)Cc2c1[2H] |
| IUPAC Name | 1,2,3,4-tetradeuterio-6-hydroxy-5,6-dihydrobenzo[b][1]benzazepine-11-carboxamide |
| Molecular Weight (g/mol) | 258.31 |
| Show More |