Learn More
10-Bromodecanoic acid, 95%
Product Code. p-7040510
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
Product Code. 15486758
Brand: Thermo Scientific Alfa Aesar H60292.06
Request Bulk or Custom Format
CAS: 50530-12-6 | C10H19BrO2 | 251.164 g/mol
It is used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is used as a pharmaceutical intermediate.
Solubility
Partly miscible in water.
Notes
Air Sensitive. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 50530-12-6
Molecular Formula C10H19BrO2
Molecular Weight (g/mol) 251.164
MDL Number MFCD00014388
InChI Key PGVRSPIEZYGOAD-UHFFFAOYSA-N
Synonym 10-bromo-decanoic acid, decanoic acid, 10-bromo, 10-bromanyldecanoic acid, 10-bromodecanoicacid, acmc-1aonn, 10-bromodecanoic acid., .omega.-bromodecanoic acid, ghl.pd_mitscher_leg0.481, ksc489q1r, 10-bromodecanoic acid
PubChem CID 142712
IUPAC Name 10-bromodecanoic acid
SMILES C(CCCCC(=O)O)CCCCBr
Specifications
Melting Point 38°C to 41°C
Linear Formula Br(CH2)9CO2H
Quantity 5 g
Solubility Information Partly miscible in water.
Formula Weight 251.16
Percent Purity 95%
Chemical Name or Material 10-Bromodecanoic acid