Learn More
Hamilton™ 10 μL 1700 Series Gas Tight Syringe
Product Code. 17814362
Click to view available options
Needle Gauge:
22s G
23s G
26S (No Needle Included)
26s G
Custom
No Needle Included
Needle Point Style:
Blunt
Cone
Curved, Beveled (No Needle Included)
Custom
No Needle Included
Sharp, Bevel, Curved, Non-coring
Connection Type:
Cemented
Removable
Unit Size:
Each
Product Code. 17814362
missing translation for 'mfr': Hamilton™ CAL80011
The 1700 Series syringes are the Gas Tight version of the 700 series syringes.
  • This series of syringes are ideal for dispensing volumes up to 10 μL.
  • A variety of different terminations and needle options are available in this series.
Specifications
Autoclavable Not Autoclavable
Material Stainless Steel
Needle Gauge 26s G
Diameter (English) Inner 0.018 in.
Diameter (English) Inner Needle 0.005 in.
Diameter (Metric) Inner Needle 0.13 mm
Diameter (English) Outer Needle 0.0187 in.
Diameter (English) Outer 0.260 in.
Diameter (Metric) Inner 0.460 mm
Diameter (Metric) Outer 6.60 mm
Diameter (Metric) Outer Needle 0.47 mm
Length (English) Needle 2 in.
Product Type Gas Tight Syringe
Max. Temperature (English) 122°F
Max. Temperature (Metric) 50°C
Min. Temperature (English) 50°F
Model 1701 N
Min. Temperature (Metric) 10°C
Quantity 1
Volume (Metric) 10 μL
Length (Metric) Needle 51 mm
Needle Point Style Sharp, Bevel, Curved, Non-coring
Connection Type Cemented